Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700030522.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030522.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030522.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030517.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030517.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030517.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030518.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030518.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030518.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030514.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030514.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030514.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030521.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030521.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030521.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030523.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030523.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030523.mm.1 |
TC0700003596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030515.mm.1 |
NM_173863.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030515.mm.1 |
NM_173863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030515.mm.1 |
TC0700003596.mm.1 |
MTA TC |