Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173863.2 | NP_776288.2 | 7 - 8 | 753 | 98 | |
GenBank mRNA | AK144840.1 | - | 7 - 8 | 754 | 98 | |
BC011210.1 | - | 7 - 8 | 592 | 4195538 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |