Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174847.2 | NP_777272.2 | 1 - 2 | 581 | 62 | |
GenBank mRNA | AK017896.1 | - | 1 - 2 | 558 | 62 | |
AK077731.1 | - | 1 - 2 | 541 | 62 | ||
AK080956.1 | - | 1 - 2 | 525 | 62 | ||
AK143875.1 | - | 1 - 2 | 555 | 62 | ||
AK220454.1 | - | 1 - 2 | 566 | 62 | ||
BC043688.1 | - | 1 - 2 | 542 | 5136661 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |