Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175022.2 | NP_778187.2 | 5 - 6 | 4322 | 145 | |
GenBank mRNA | AK032894.1 | - | 2 - 3 | 809 | 146 | |
AK173130.1 | - | 2 - 3 | 3761 | 145 | ||
BC058674.1 | - | 2 - 3 | 2961 | 145 | ||
BC075628.1 | - | 6 - 7 | 3231 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |