Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175022.2 | NP_778187.2 | 7 - 8 | 4875 | 73 | |
GenBank mRNA | AK032894.1 | - | 4 - 5 | 1363 | 73 | |
AK173130.1 | - | 4 - 5 | 4314 | 73 | ||
BC058674.1 | - | 4 - 5 | 3514 | 73 | ||
BC075628.1 | - | 8 - 9 | 3784 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |