Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175259.4 | NP_780468.1 | 2 - 3 | 345 | 59 | |
GenBank mRNA | AK039457.1 | - | 2 - 3 | 356 | 59 | |
BC044813.1 | - | 2 - 3 | 345 | 4950177 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |