Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176860.5 | NP_789830.1 | 11 - 12 | 1630 | 67 | |
GenBank mRNA | AB075602.1 | - | 11 - 12 | 1625 | 67 | |
AK013361.1 | - | 11 - 12 | 1698 | 67 | ||
AK034450.1 | - | 11 - 12 | 1821 | 67 | ||
AK035764.1 | - | 11 - 12 | 1631 | 67 | ||
AK133948.1 | - | 11 - 12 | 1628 | 67 | ||
AK150909.1 | - | 11 - 12 | 2197 | 67 | ||
AK151721.1 | - | 11 - 12 | 2198 | 67 | ||
AK151895.1 | - | 11 - 12 | 1831 | 67 | ||
AK152013.1 | - | 11 - 12 | 1831 | 67 | ||
AK154576.1 | - | 11 - 12 | 1852 | 67 | ||
BC053436.1 | - | 11 - 12 | 1602 | 6510702 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |