Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164785.1 | NP_001158257.1 | 6 - 7 | 1326 | 75 | |
NM_001164786.1 | NP_001158258.1 | 6 - 7 | 1326 | 75 | ||
NM_177431.4 | NP_803180.3 | 6 - 7 | 1326 | 75 | ||
GenBank mRNA | AJ512753.3 | - | 6 - 7 | 1095 | 75 | |
AK090358.1 | - | 6 - 7 | 1292 | 75 | ||
AY189815.1 | - | 6 - 7 | 1234 | 75 | ||
AY189816.1 | - | 6 - 7 | 1234 | 75 | ||
BC121792.1 | - | 6 - 7 | 1165 | 40099393 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |