Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177664.5 | NP_808332.3 | 5 - 6 | 1416 | 72 | |
GenBank mRNA | AF541279.1 | - | 5 - 6 | 1417 | 72 | |
AK046782.1 | - | 5 - 6 | 1417 | 72 | ||
AK079635.1 | - | 5 - 6 | 750 | 72 | ||
AK129149.1 | - | 5 - 6 | 1012 | 72 | ||
AY266266.1 | - | 5 - 6 | 1417 | 72 | ||
AY266267.1 | - | 5 - 6 | 1417 | 72 | ||
BC082323.1 | - | 4 - 5 | 2394 | 72 | ||
BC137701.1 | - | 5 - 6 | 1011 | 8860720 | 72 | |
BC137702.1 | - | 5 - 6 | 1012 | 8860721 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |