Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178016.3 | NP_821135.1 | 6 - 7 | 666 | 124 | |
GenBank mRNA | AK046246.1 | - | 6 - 7 | 654 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |