Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178617.4 | NP_848732.3 | 11 - 12 | 1157 | 66 | |
GenBank mRNA | AK039240.1 | - | 11 - 12 | 973 | 66 | |
AK050307.1 | - | 12 - 13 | 1054 | 66 | ||
AK081951.1 | - | 11 - 12 | 1049 | 66 | ||
AY278200.2 | - | 11 - 12 | 1140 | 66 | ||
BC067055.1 | - | 11 - 12 | 1157 | 30545678 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |