Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178617.4 | NP_848732.3 | 3 - 4 | 444 | 105 | |
GenBank mRNA | AK039240.1 | - | 3 - 4 | 260 | 105 | |
AK050307.1 | - | 4 - 5 | 341 | 105 | ||
AK081951.1 | - | 3 - 4 | 336 | 105 | ||
AY278200.2 | - | 3 - 4 | 427 | 105 | ||
BC067055.1 | - | 3 - 4 | 444 | 30545678 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |