Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178635.3 | NP_848750.3 | 9 - 10 | 1104 | 70 | |
GenBank mRNA | AB020334.1 | - | 8 - 9 | 709 | 70 | |
AK030814.1 | - | 9 - 10 | 1035 | 70 | ||
AK043084.1 | - | 9 - 10 | 1082 | 70 | ||
AK044129.1 | - | 10 - 11 | 1200 | 70 | ||
AK078085.1 | - | 9 - 10 | 1096 | 70 | ||
AK089900.1 | - | 9 - 10 | 1089 | 70 | ||
AK154903.1 | - | 9 - 10 | 1104 | 70 | ||
AK165015.1 | - | 9 - 10 | 1094 | 70 | ||
AK165185.1 | - | 9 - 10 | 1084 | 70 | ||
BC034072.1 | - | 3 - 4 | 290 | 70 | ||
BC034176.1 | - | 9 - 10 | 1073 | 4954871 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |