Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199247.1 | NP_001186176.1 | 11 - 12 | 1232 | 142 | |
NM_001199248.1 | NP_001186177.1 | 10 - 11 | 1331 | 142 | ||
NM_178637.2 | NP_848752.1 | 12 - 13 | 1388 | 142 | ||
NR_037603.1 | 9 - 10 | 1264 | 142 | |||
GenBank mRNA | AB055409.1 | - | 9 - 10 | 1242 | 142 | |
AF528194.1 | - | 8 - 9 | 942 | 142 | ||
AF528195.1 | - | 11 - 12 | 1323 | 142 | ||
AF528196.1 | - | 10 - 11 | 1167 | 142 | ||
AK146352.1 | - | 12 - 13 | 1345 | 142 | ||
AK159568.1 | - | 10 - 11 | 1264 | 142 | ||
AK160077.1 | - | 10 - 11 | 1264 | 142 | ||
BC022768.1 | - | 3 - 4 | 313 | 142 | ||
BC110675.1 | - | 13 - 14 | 1622 | 4011689 | 142 | |
BC129967.1 | - | 10 - 11 | 1166 | 142 | ||
BC129968.1 | - | 10 - 11 | 1167 | 40106268 | 142 | |
DQ139980.1 | - | 12 - 13 | 1616 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |