Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242349.1 | NP_001229278.1 | 6 - 7 | 1015 | 91 | |
NM_178642.5 | NP_848757.4 | 6 - 7 | 1012 | 91 | ||
GenBank mRNA | AK052589.1 | - | 6 - 7 | 964 | 91 | |
BC062959.1 | - | 6 - 7 | 901 | 30547439 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |