Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178746.4 | NP_848861.1 | 13 - 14 | 1569 | 86 | |
GenBank mRNA | AK043402.1 | - | 13 - 14 | 1557 | 86 | |
AK044735.1 | - | 13 - 14 | 1559 | 86 | ||
AK078926.1 | - | 7 - 8 | 809 | 86 | ||
BC052361.1 | - | 13 - 14 | 6705789 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |