Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178899.5 | NP_849230.1 | 4 - 5 | 1032 | 71 | |
GenBank mRNA | AK033728.1 | - | 4 - 5 | 1034 | 71 | |
AK050501.1 | - | 4 - 5 | 1033 | 71 | ||
BC096374.1 | - | 4 - 5 | 1019 | 4483627 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |