Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178909.4 | NP_849240.1 | 5 - 6 | 705 | 108 | |
GenBank mRNA | AK033294.1 | - | 5 - 6 | 651 | 108 | |
AK047691.1 | - | 5 - 6 | 679 | 108 | ||
AK088218.1 | - | 5 - 6 | 680 | 108 | ||
AK142228.1 | - | 5 - 6 | 651 | 108 | ||
BC096540.1 | - | 5 - 6 | 634 | 6403681 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |