Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181848.4 | NP_862896.1 | 10 - 11 | 1291 | 100 | |
GenBank mRNA | AY071834.1 | - | 10 - 11 | 1398 | 100 | |
BC024755.1 | - | 3 - 4 | 1177 | 100 | ||
BC061185.1 | - | 10 - 11 | 1291 | 30314331 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |