Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199257.2 | NP_954866.2 | 2 - 3 | 436 | 78 | |
GenBank mRNA | AJ311311.1 | - | 2 - 3 | 439 | 78 | |
AJ311312.1 | - | 2 - 3 | 439 | 78 | ||
AJ311313.1 | - | 2 - 3 | 439 | 78 | ||
BC107329.2 | - | 2 - 3 | 231 | 40054415 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |