Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
366437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229016 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
710328 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
117778 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521199 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821663 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
888870 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
84252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364713 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
967476 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898169 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
738197 |
NM_001005569.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
134091 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
280 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139151 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P145530 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176053 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176046 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176052 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176031 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176050 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176033 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176058 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176036 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176035 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176057 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176060 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176055 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31176045 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P145530 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139151 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03315510 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
M92849.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
NM_009638.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
AK009449.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
BC011150.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P370268 |
L05559.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130645 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P145530 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139151 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13401334 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13301355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31175938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31176050 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13305753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176020 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13440987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176022 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176049 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176052 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176057 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176033 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13175969 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176068 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13401334 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176012 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176024 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176019 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176045 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176065 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176044 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13526307 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176055 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176054 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176030 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176059 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13154606 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176050 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176048 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31176060 |
- |
- |