Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049519.1 | - | 8 - 9 | 1605 | 103 | |
AK081690.1 | - | 8 - 9 | 1576 | 103 | ||
BC017547.1 | - | 8 - 9 | 1427 | 4920702 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |