Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013776.1 | - | 4 | 1554 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |