Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 30 - 31 | 2095 | 74 | |
GenBank mRNA | AK132180.1 | - | 30 - 31 | 2115 | 74 | |
AK158011.1 | - | 30 - 31 | 2112 | 74 | ||
AK159283.1 | - | 30 - 31 | 2113 | 74 | ||
AK159285.1 | - | 30 - 31 | 2115 | 74 | ||
BC042642.1 | - | 23 - 24 | 1665 | 74 | ||
BC050014.1 | - | 30 - 31 | 2095 | 4225038 | 74 | |
BC059281.1 | - | 30 - 31 | 2105 | 3992363 | 74 | |
M14423.1 | - | 8 - 9 | 445 | 74 | ||
U08020.1 | - | 30 - 31 | 1996 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |