Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 37 - 38 | 2685 | 152 | |
GenBank mRNA | AK132180.1 | - | 37 - 38 | 2706 | 152 | |
AK159283.1 | - | 37 - 38 | 2703 | 152 | ||
AK159285.1 | - | 37 - 38 | 2706 | 153 | ||
BC042642.1 | - | 30 - 31 | 2255 | 152 | ||
BC050014.1 | - | 37 - 38 | 2685 | 4225038 | 152 | |
M14423.1 | - | 15 - 16 | 1035 | 152 | ||
U08020.1 | - | 37 - 38 | 2586 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |