Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008296.2 | NP_032322.1 | 8 - 9 | 1012 | 80 | |
GenBank mRNA | AK075651.1 | - | 4 - 5 | 787 | 80 | |
AK089144.1 | - | 7 - 8 | 915 | 80 | ||
AK214801.1 | - | 1 - 2 | 53 | 80 | ||
BC013716.1 | - | 8 - 9 | 1012 | 3978881 | 80 | |
BC094064.1 | - | 8 - 9 | 1022 | 4158683 | 80 | |
KM043052.1 | - | 8 - 9 | 1012 | 80 | ||
KM043053.1 | - | 8 - 9 | 1012 | 80 | ||
KM043054.1 | - | 8 - 9 | 1012 | 80 | ||
KM043055.1 | - | 8 - 9 | 1012 | 80 | ||
X61753.1 | - | 8 - 9 | 995 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |