Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281819.1 | NP_001268748.1 | 21 - 22 | 3337 | 99 | |
NM_009598.2 | NP_033728.1 | 10 - 11 | 1560 | 99 | ||
NM_207624.5 | NP_997507.1 | 21 - 22 | 3337 | 99 | ||
GenBank mRNA | AK154632.1 | - | 21 - 22 | 3336 | 99 | |
AK161020.1 | - | 21 - 22 | 3333 | 99 | ||
AK164780.1 | - | 8 - 9 | 1205 | 99 | ||
BC034367.1 | - | 17 - 18 | 2599 | 99 | ||
BC040404.1 | - | 21 - 22 | 3326 | 3965649 | 99 | |
BC083109.1 | - | 21 - 22 | 3115 | 99 | ||
BC110362.1 | - | 21 - 22 | 3323 | 5343978 | 99 | |
J04946.1 | - | 15 - 16 | 2304 | 99 | ||
J04947.1 | - | 2304 | 99 | |||
M55333.1 | - | 10 - 11 | 1560 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |