Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201413.1 | NP_001188342.1 | 13 - 14 | 2297 | 64 | |
NM_001201414.1 | NP_001188343.1 | 12 - 13 | 2300 | 64 | ||
NM_001201415.1 | NP_001188344.1 | 12 - 13 | 2234 | 64 | ||
NM_001201416.1 | NP_001188345.1 | 3 - 4 | 400 | 64 | ||
NM_009686.2 | NP_033816.1 | 14 - 15 | 2303 | 64 | ||
GenBank mRNA | AK004792.1 | - | 14 - 15 | 2157 | 64 | |
AK153260.1 | - | 12 - 13 | 2266 | 64 | ||
AK167649.1 | - | 13 - 14 | 2162 | 64 | ||
AK172405.1 | - | 3 - 4 | 404 | 64 | ||
BC042797.1 | - | 4 - 5 | 442 | 64 | ||
BC076587.1 | - | 12 - 13 | 2080 | 30550087 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |