Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243052.1 | NP_001229981.1 | 9 - 10 | 1320 | 59 | |
NM_001243053.1 | NP_001229982.1 | 10 - 11 | 1205 | 59 | ||
NM_009737.3 | NP_033867.1 | 9 - 10 | 1293 | 59 | ||
GenBank mRNA | AF031467.1 | - | 9 - 10 | 1085 | 59 | |
AK137049.1 | - | 10 - 11 | 1256 | 59 | ||
AK145300.1 | - | 9 - 10 | 1138 | 59 | ||
AK160159.1 | - | 9 - 10 | 1321 | 59 | ||
BC017688.1 | - | 9 - 10 | 1070 | 4242224 | 59 | |
BC048072.1 | - | 9 - 10 | 1084 | 4166264 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |