Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010194.2 | NP_034324.2 | 12 - 13 | 1774 | 65 | |
GenBank mRNA | AK004587.1 | - | 11 - 12 | 1902 | 65 | |
AK143639.1 | - | 12 - 13 | 1724 | 65 | ||
AK155861.1 | - | 10 - 11 | 2148 | 65 | ||
AK170418.1 | - | 12 - 13 | 1715 | 65 | ||
BC100306.1 | - | 11 - 12 | 1907 | 65 | ||
BC129919.1 | - | 11 - 12 | 1710 | 40111855 | 65 | |
X12616.1 | - | 11 - 12 | 1708 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |