Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010295.2 | NP_034425.1 | 11 - 12 | 1566 | 90 | |
GenBank mRNA | AK144339.1 | - | 11 - 12 | 1567 | 90 | |
AK160876.1 | - | 11 - 12 | 1326 | 90 | ||
AK169676.1 | - | 11 - 12 | 1539 | 90 | ||
BC019374.1 | - | 11 - 12 | 1323 | 4195425 | 90 | |
U85414.1 | - | 11 - 12 | 1291 | 90 | ||
U85498.1 | - | 11 - 12 | 1291 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |