Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010306.3 | NP_034436.1 | 4 - 5 | 605 | 91 | |
GenBank mRNA | AK004566.1 | - | 4 - 5 | 599 | 91 | |
AK077490.1 | - | 4 - 5 | 567 | 91 | ||
AK133749.1 | - | 4 - 5 | 546 | 91 | ||
AK167438.1 | - | 4 - 5 | 559 | 91 | ||
AK168578.1 | - | 4 - 5 | 543 | 91 | ||
AK169067.1 | - | 4 - 5 | 550 | 91 | ||
AK169285.1 | - | 4 - 5 | 580 | 91 | ||
BC003462.1 | - | 4 - 5 | 531 | 91 | ||
BC020009.1 | - | 4 - 5 | 577 | 91 | ||
BC041107.1 | - | 4 - 5 | 516 | 2648164 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |