Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010472.2 | NP_034602.1 | 7 - 8 | 1259 | 70 | |
GenBank mRNA | AF057287.1 | - | 2 - 3 | 317 | 70 | |
AK048326.1 | - | 7 - 8 | 1260 | 70 | ||
AK049400.1 | - | 7 - 8 | 1246 | 70 | ||
BC031154.1 | - | 7 - 8 | 1246 | 4976622 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |