Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 6 - 7 | 2067 | 68 | |
GenBank mRNA | AF056187.1 | - | 6 - 7 | 1465 | 68 | |
AK137220.1 | - | 1 - 2 | 196 | 68 | ||
AK155079.1 | - | 5 - 6 | 945 | 68 | ||
AK155884.1 | - | 5 - 6 | 963 | 68 | ||
BC138868.1 | - | 6 - 7 | 1497 | 8861890 | 68 | |
BC138869.1 | - | 6 - 7 | 1497 | 8861891 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |