Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010616.2 | NP_034746.1 | 6 - 7 | 621 | 60 | |
GenBank mRNA | AB001428.1 | - | 2 - 3 | 198 | 60 | |
AB755406.1 | - | 6 - 7 | 634 | 60 | ||
AK018598.1 | - | 6 - 7 | 623 | 60 | ||
BC022225.1 | - | 6 - 7 | 551 | 4222522 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |