Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010713.2 | NP_034843.2 | 5 - 6 | 874 | 64 | |
GenBank mRNA | AJ000338.1 | - | 5 - 6 | 974 | 64 | |
AK160121.1 | - | 5 - 6 | 875 | 64 | ||
AK165733.1 | - | 5 - 6 | 868 | 64 | ||
BC125281.1 | - | 5 - 6 | 783 | 40129796 | 64 | |
BC125283.1 | - | 5 - 6 | 783 | 40129798 | 64 | |
BC144768.1 | - | 5 - 6 | 783 | 9053316 | 64 | |
D49658.1 | - | 5 - 6 | 843 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |