Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139516.1 | NP_001132988.1 | 10 - 11 | 1434 | 125 | |
NM_011249.2 | NP_035379.2 | 10 - 11 | 1434 | 125 | ||
GenBank mRNA | AK033402.1 | - | 10 - 11 | 1428 | 125 | |
AK147064.1 | - | 10 - 11 | 1435 | 125 | ||
AK156055.1 | - | 10 - 11 | 1413 | 125 | ||
AK214366.1 | - | 1 - 2 | 126 | 125 | ||
BC060124.1 | - | 10 - 11 | 1401 | 6408844 | 125 | |
BC069179.1 | - | 2 - 3 | 212 | 125 | ||
U27177.1 | - | 10 - 11 | 1418 | 125 | ||
U27178.1 | - | 10 - 11 | 1418 | 125 | ||
U33320.1 | - | 10 - 11 | 1401 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |