Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204979.1 | NP_001191908.1 | 2 - 3 | 337 | 118 | |
NM_011319.3 | NP_035449.1 | 2 - 3 | 337 | 118 | ||
GenBank mRNA | AK028358.1 | - | 2 - 3 | 269 | 118 | |
AK075827.1 | - | 2 - 3 | 268 | 118 | ||
AK077309.1 | - | 2 - 3 | 219 | 118 | ||
AK082793.1 | - | 2 - 3 | 248 | 118 | ||
AK147825.1 | - | 2 - 3 | 236 | 118 | ||
AK152150.1 | - | 2 - 3 | 295 | 118 | ||
AK153158.1 | - | 2 - 3 | 287 | 118 | ||
BC008612.1 | - | 2 - 3 | 226 | 3257101 | 118 | |
BC031932.1 | - | 2 - 3 | 226 | 118 | ||
M74012.1 | - | 2 - 3 | 289 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |