Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011947.3 | NP_036077.1 | 9 - 10 | 805 | 77 | |
GenBank mRNA | AK137127.1 | - | 9 - 10 | 1091 | 77 | |
AK220450.1 | - | 9 - 10 | 1099 | 77 | ||
BC023781.1 | - | 9 - 10 | 809 | 5346982 | 77 | |
U43187.1 | - | 9 - 10 | 1108 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |