Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011965.2 | NP_036095.1 | 4 - 5 | 365 | 85 | |
GenBank mRNA | AF060088.1 | - | 4 - 5 | 289 | 85 | |
AJ272019.1 | - | 4 - 5 | 318 | 85 | ||
AJ272020.1 | - | 1 - 2 | 35 | 85 | ||
AK088673.1 | - | 4 - 5 | 332 | 85 | ||
AK135326.1 | - | 4 - 5 | 303 | 85 | ||
AK137054.1 | - | 4 - 5 | 307 | 85 | ||
AK144784.1 | - | 4 - 5 | 320 | 85 | ||
AK162277.1 | - | 4 - 5 | 312 | 85 | ||
AK167689.1 | - | 4 - 5 | 303 | 85 | ||
BC005762.1 | - | 4 - 5 | 302 | 2655483 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |