Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012037.2 | NP_036167.1 | 2 - 3 | 703 | 94 | |
GenBank mRNA | AK034795.1 | - | 2 - 3 | 705 | 94 | |
AK145193.1 | - | 2 - 3 | 737 | 94 | ||
AK154178.1 | - | 688 | 94 | |||
AK154965.1 | - | 2 - 3 | 686 | 94 | ||
AK155342.1 | - | 2 - 3 | 704 | 94 | ||
AK159310.1 | - | 2 - 3 | 646 | 94 | ||
AK159315.1 | - | 2 - 3 | 688 | 94 | ||
AK160891.1 | - | 2 - 3 | 695 | 94 | ||
AK170371.1 | - | 2 - 3 | 684 | 94 | ||
AK194936.1 | - | 1 - 2 | 184 | 94 | ||
BC025496.1 | - | 2 - 3 | 659 | 5320239 | 94 | |
BC036169.1 | - | 1 - 2 | 145 | 94 | ||
BC085504.1 | - | 2 - 3 | 622 | 94 | ||
BC099678.1 | - | 2 - 3 | 685 | 6311681 | 94 | |
X95562.1 | - | 2 - 3 | 325 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |