Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285870.1 | NP_001272799.1 | 4 - 5 | 744 | 61 | |
NM_001285871.1 | NP_001272800.1 | 4 - 5 | 929 | 61 | ||
NM_001285873.1 | NP_001272802.1 | 4 - 5 | 744 | 61 | ||
NM_018880.3 | NP_061368.1 | 4 - 5 | 718 | 61 | ||
GenBank mRNA | AB030912.1 | - | 4 - 5 | 745 | 61 | |
AF220019.1 | - | 4 - 5 | 637 | 61 | ||
AK019165.1 | - | 4 - 5 | 679 | 61 | ||
AK144674.1 | - | 4 - 5 | 647 | 61 | ||
AK170029.1 | - | 4 - 5 | 682 | 61 | ||
BC003347.1 | - | 4 - 5 | 759 | 61 | ||
BC034263.1 | - | 4 - 5 | 683 | 4503884 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |