Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020333.2 | NP_065066.2 | 20 - 21 | 3094 | 131 | |
GenBank mRNA | AF332063.1 | - | 20 - 21 | 2764 | 131 | |
AF332064.1 | - | 20 - 21 | 2764 | 131 | ||
AK122460.1 | - | 20 - 21 | 2828 | 131 | ||
AK147262.1 | - | 20 - 21 | 3096 | 131 | ||
BC054808.1 | - | 20 - 21 | 2760 | 6838880 | 131 | |
BC057624.1 | - | 1 - 2 | 144 | 131 | ||
KJ535320.1 | - | 20 - 21 | 2785 | 131 | ||
KJ535321.1 | - | 20 - 21 | 2785 | 131 | ||
KJ535322.1 | - | 20 - 21 | 2941 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |