Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022416.2 | NP_071861.1 | 8 - 9 | 1157 | 97 | |
GenBank mRNA | AJ250840.1 | - | 8 - 9 | 1235 | 97 | |
AK082468.1 | - | 8 - 9 | 1158 | 97 | ||
BC052404.1 | - | 8 - 9 | 1067 | 5715729 | 97 | |
BC056396.1 | - | 8 - 9 | 1143 | 6841272 | 97 | |
BC058412.1 | - | 8 - 9 | 1067 | 5715729 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |