Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026236.3 | NP_080512.1 | 11 - 12 | 1183 | 104 | |
GenBank mRNA | AK012599.1 | - | 11 - 12 | 1183 | 104 | |
AK018396.1 | - | 11 - 12 | 1186 | 104 | ||
AK030041.1 | - | 11 - 12 | 1184 | 104 | ||
AK031244.1 | - | 11 - 12 | 1196 | 104 | ||
AK033631.1 | - | 11 - 12 | 1182 | 104 | ||
AK035273.1 | - | 11 - 12 | 1185 | 104 | ||
AK035864.1 | - | 11 - 12 | 1185 | 104 | ||
AK043970.1 | - | 11 - 12 | 1185 | 104 | ||
AK122511.1 | - | 10 - 11 | 2325 | 104 | ||
AK128953.1 | - | 11 - 12 | 1183 | 104 | ||
BC006679.1 | - | 10 - 11 | 1046 | 104 | ||
BC021348.1 | - | 1 - 2 | 1373 | 104 | ||
BC040795.1 | - | 7 - 8 | 714 | 104 | ||
BC048155.1 | - | 11 - 12 | 1172 | 5698339 | 104 | |
BC050106.1 | - | 11 - 12 | 1200 | 104 | ||
BC062967.1 | - | 11 - 12 | 1187 | 6835444 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |