Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026236.3 | NP_080512.1 | 2 - 3 | 204 | 104 | |
GenBank mRNA | AK012599.1 | - | 2 - 3 | 204 | 104 | |
AK018396.1 | - | 2 - 3 | 207 | 104 | ||
AK030041.1 | - | 2 - 3 | 205 | 104 | ||
AK031244.1 | - | 2 - 3 | 105 | |||
AK033631.1 | - | 2 - 3 | 203 | 104 | ||
AK035273.1 | - | 2 - 3 | 206 | 104 | ||
AK035864.1 | - | 2 - 3 | 206 | 104 | ||
AK043970.1 | - | 2 - 3 | 206 | 104 | ||
AK122511.1 | - | 2 - 3 | 188 | 104 | ||
AK128953.1 | - | 2 - 3 | 204 | 104 | ||
BC006679.1 | - | 1 - 2 | 67 | 104 | ||
BC048155.1 | - | 2 - 3 | 193 | 5698339 | 104 | |
BC050106.1 | - | 2 - 3 | 221 | 104 | ||
BC062967.1 | - | 2 - 3 | 208 | 6835444 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |