Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026460.3 | NP_080736.2 | 7 - 8 | 1107 | 101 | |
GenBank mRNA | AF251276.1 | - | 7 - 8 | 1092 | 101 | |
AK007347.1 | - | 7 - 8 | 1108 | 101 | ||
AK007510.1 | - | 7 - 8 | 1082 | 101 | ||
BC098466.1 | - | 7 - 8 | 1084 | 6433250 | 101 | |
CF581490.1 | - | 2 - 3 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |