Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026791.2 | NP_081067.2 | 2 - 3 | 561 | 57 | |
GenBank mRNA | AK082085.1 | - | 2 - 3 | 965 | 57 | |
AK156709.1 | - | 2 - 3 | 627 | 57 | ||
BB870341.1 | - | 2 - 3 | 172 | 57 | ||
BC043658.1 | - | 2 - 3 | 522 | 57 | ||
BC144773.1 | - | 2 - 3 | 927 | 57 | ||
BC145661.1 | - | 2 - 3 | 927 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |