Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242358.1 | NP_001229287.1 | 5 - 6 | 755 | 100 | |
NM_027296.3 | NP_081572.1 | 5 - 6 | 690 | 100 | ||
GenBank mRNA | AK011764.1 | - | 5 - 6 | 706 | 100 | |
AK139764.1 | - | 5 - 6 | 672 | 100 | ||
AK152739.1 | - | 5 - 6 | 726 | 100 | ||
AK166804.1 | - | 5 - 6 | 700 | 100 | ||
BC031764.1 | - | 5 - 6 | 656 | 4486420 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |