Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028312.3 | NP_082588.2 | 4 - 5 | 363 | 76 | |
GenBank mRNA | AK012610.1 | - | 4 - 5 | 364 | 76 | |
BC026668.1 | - | 4 - 5 | 313 | 5006213 | 76 | |
CA450868.1 | - | 2 - 3 | 97 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |